Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012120.4 | XP_004012169.2 | 7 - 8 | 1774 | 82 | |
XM_015098118.2 | XP_014953604.2 | 7 - 8 | 1759 | 82 | ||
XM_027973610.1 | XP_027829411.1 | 7 - 8 | 1706 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
