Assay Details
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022247.4 | XP_004022296.1 | 1 - 2 | 311 | 78 | |
XM_012107046.3 | XP_011962436.1 | 1 - 2 | 312 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
