Assay Details
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162547.1 | NP_001156019.1 | 5 - 6 | 453 | 104 | |
GenBank mRNA | FJ969399.1 | - | 5 - 6 | 453 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
