Assay Details
Gene Symbol: LOC101108212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967845.1 | XP_027823646.1 | 2 - 3 | 864 | 75 |
Gene Symbol: LOC114114054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967844.1 | XP_027823645.1 | 2 - 3 | 268 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
