Assay Details
Gene Symbol: RBBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020479.4 | XP_004020528.2 | 13 - 14 | 2294 | 126 | |
XM_012103494.3 | XP_011958884.2 | 12 - 13 | 2163 | 126 |
Gene Symbol: LOC101121879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096517.2 | XP_011951907.2 | 1 | 1914 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
