Assay Details
Gene Symbol: LOC101120386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004709.4 | XP_004004758.1 | 4 - 5 | 759 | 89 |
Gene Symbol: LOC114110469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960941.1 | XP_027816742.1 | 3 - 4 | 376 | 89 |
Gene Symbol: LOC114113132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964817.1 | XP_027820618.1 | 3 - 4 | 439 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
