Assay Details
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980298.1 | XP_027836099.1 | 2 - 3 | 662 | 114 | |
XM_027980303.1 | XP_027836104.1 | 2 - 3 | 632 | 114 | ||
XM_027980311.1 | XP_027836112.1 | 2 - 3 | 623 | 114 | ||
XM_027980320.1 | XP_027836121.1 | 2 - 3 | 837 | 114 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
