Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095934.2 | XP_014951420.1 | 4 - 5 | 692 | 95 | |
XM_015095935.2 | XP_014951421.1 | 5 - 6 | 749 | 95 | ||
XM_015095936.2 | XP_014951422.1 | 3 - 4 | 510 | 95 | ||
XM_027969959.1 | XP_027825760.1 | 4 - 5 | 602 | 95 |
