Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012133548.3 | XP_011988938.1 | 9 - 10 | 1041 | 94 | |
XM_027964843.1 | XP_027820644.1 | 9 - 10 | 938 | 94 | ||
XM_027964844.1 | XP_027820645.1 | 9 - 10 | 823 | 94 | ||
XM_027964845.1 | XP_027820646.1 | 9 - 10 | 895 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
