Assay Details
Gene Symbol: MECP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963279.1 | XP_027819080.1 | 2 - 3 | 526 | 87 | |
XM_027963280.1 | XP_027819081.1 | 3 - 4 | 596 | 87 | ||
XM_027963281.1 | XP_027819082.1 | 2 - 3 | 1516 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
