Assay Details
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012118654.3 | XP_011974044.2 | 6 - 7 | 2234 | 54 | |
XM_027964311.1 | XP_027820112.1 | 6 - 7 | 2114 | 54 | ||
XM_027964312.1 | XP_027820113.1 | 2 - 3 | 952 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
