Assay Details
Gene Symbol: MYCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973699.1 | XP_027829500.1 | 25 - 26 | 3969 | 104 |
Gene Symbol: LOC114116662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973851.1 | XP_027829652.1 | 7 - 8 | 845 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
