Assay Details
Gene Symbol: LOC114108792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956275.1 | XP_027812076.1 | 1 | 1365 | 119 |
Gene Symbol: ADSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976332.1 | XP_027832133.1 | 12 - 13 | 1415 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
