Assay Details
Gene Symbol: PPP4R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005937.4 | XP_004005986.3 | 5 - 6 | 1204 | 70 | |
XM_015094378.2 | XP_014949864.2 | 5 - 6 | 1204 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
