Assay Details
Gene Symbol: ITPRID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004533.4 | XP_004004582.3 | 15 - 16 | 3230 | 83 | |
XM_027964773.1 | XP_027820574.1 | 15 - 16 | 3227 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
