Assay Details
Gene Symbol: LOC114117326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976155.1 | XP_027831956.1 | 2 | 1265 | 67 |
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971185.1 | XP_027826986.1 | 20 - 21 | 8483 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
