Assay Details
Gene Symbol: MRTFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962081.1 | XP_027817882.1 | 7 - 8 | 702 | 76 | |
XM_027962082.1 | XP_027817883.1 | 5 - 6 | 467 | 76 | ||
XM_027962083.1 | XP_027817884.1 | 5 - 6 | 611 | 76 | ||
XM_027962084.1 | XP_027817885.1 | 5 - 6 | 442 | 76 | ||
XM_027962085.1 | XP_027817886.1 | 3 - 4 | 1711 | 76 | ||
XM_027962086.1 | XP_027817887.1 | 7 - 8 | 702 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
