Assay Details
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973107.1 | XP_027828908.1 | 4 - 5 | 548 | 61 | |
XM_027973108.1 | XP_027828909.1 | 3 - 4 | 491 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
