Assay Details
Gene Symbol: LOC101122444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011715.4 | XP_004011764.2 | 5 - 6 | 534 | 101 | |
XM_012183791.3 | XP_012039181.1 | 5 - 6 | 520 | 101 | ||
XM_012183794.3 | XP_012039184.1 | 4 - 5 | 450 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
