Assay Details
Gene Symbol: MIR218A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107959.1 | 1 | 32 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | slit guidance ligand 3 | SLIT3 | Sense | Chr. 16 - 617590 - 1388004 on NCBI Build Oar_rambouillet_v1.0 | ||
Downstream | spindle apparatus coiled-coil protein 1 | SPDL1 | 986.2kb | Anti-sense | Chr. 16 - 1710495 - 1743689 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
