Assay Details
Gene Symbol: MIR10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107943.1 | 1 | 42 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox B3 | HOXB3 | -3.6kb | Sense | Chr. 11 - 25386736 - 25411776 on NCBI Build Oar_rambouillet_v1.0 | |
Downstream | homeobox B5 | HOXB5 |
HOX-2.1, OHox-2.1 |
11.1kb | Sense | Chr. 11 - 25426579 - 25430985 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
