Assay Details
Gene Symbol: CYP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123000.1 | NP_001116472.1 | 6 - 7 | 854 | 103 | |
GenBank mRNA | AJ012153.1 | - | 6 - 7 | 886 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
