Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009806.1 | NP_001009806.1 | 2 - 3 | 207 | 126 | |
GenBank mRNA | X53934.1 | - | 207 | 126 | ||
X55641.1 | - | 207 | 126 | |||
X60148.1 | - | 147 | 126 |

More Information
Set Membership: |
![]() ![]() |
