Assay Details
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145177.1 | NP_001138649.1 | 2 - 3 | 534 | 86 | |
GenBank mRNA | FJ589639.1 | - | 2 - 3 | 534 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
