Assay Details
Gene Symbol: IGFBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159276.1 | NP_001152748.1 | 3 - 4 | 764 | 87 | |
XM_027968353.1 | XP_027824154.1 | 3 - 4 | 872 | 87 | ||
GenBank mRNA | AF327651.1 | - | 2 - 3 | 264 | 87 | |
EU616622.1 | - | 3 - 4 | 792 | 87 | ||
FJ752574.1 | - | 3 - 4 | 764 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
