Assay Details
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040288.2 | NP_001035378.1 | 2 - 3 | 438 | 76 | |
GenBank mRNA | DQ228866.1 | - | 2 - 3 | 442 | 76 | |
MG669627.1 | - | 2 - 3 | 420 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
