Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024860.1 | NP_001020031.1 | 1 - 2 | 355 | 84 | |
GenBank mRNA | X55152.1 | - | 339 | 84 | ||
X56756.1 | - | 355 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
