Assay Details
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009436.1 | NP_001009436.1 | 1 - 2 | 472 | 95 | |
GenBank mRNA | S44612.1 | - | 1 - 2 | 472 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
