Assay Details
Gene Symbol: TIMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166187.1 | NP_001159659.1 | 2 - 3 | 206 | 94 | |
GenBank mRNA | GQ221039.1 | - | 2 - 3 | 206 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
