Assay Details
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199795.1 | NP_001186724.1 | 6 - 7 | 640 | 81 | |
GenBank mRNA | HM628580.1 | - | 6 - 7 | 640 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
