Assay Details
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009191.1 | NP_001009191.1 | 3 - 4 | 331 | 96 | |
GenBank mRNA | AB070717.1 | - | 3 - 4 | 331 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
