Assay Details
Gene Symbol: LIN28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256768.1 | NP_001243697.1 | 3 - 4 | 415 | 74 | |
XM_015097249.2 | XP_014952735.1 | 2 - 3 | 378 | 74 | ||
GenBank mRNA | JQ277701.1 | - | 3 - 4 | 415 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
