Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166182.1 | NP_001159654.1 | 1 - 2 | 262 | 77 | |
GenBank mRNA | GQ221038.1 | - | 1 - 2 | 262 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
