Assay Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166182.1 | NP_001159654.1 | 2 - 3 | 516 | 104 | |
GenBank mRNA | GQ221038.1 | - | 2 - 3 | 516 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
