Assay Details
Gene Symbol: VAMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166204.1 | NP_001159676.1 | 5 - 6 | 506 | 95 | |
XM_012106120.3 | XP_011961510.1 | 6 - 7 | 649 | 95 | ||
GenBank mRNA | GQ221059.1 | - | 5 - 6 | 506 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
