Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082594.1 | NP_001076063.1 | 1 - 2 | 187 | 83 | |
GenBank mRNA | DQ679798.1 | - | 1 - 2 | 187 | 83 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082594.1 | NP_001076063.1 | 1 - 2 | 187 | 83 | |
GenBank mRNA | DQ679798.1 | - | 1 - 2 | 187 | 83 |