Assay Details
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145177.1 | NP_001138649.1 | 3 - 4 | 667 | 93 | |
GenBank mRNA | FJ589639.1 | - | 3 - 4 | 667 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
