Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024860.1 | NP_001020031.1 | 2 - 3 | 410 | 60 | |
XM_012100437.3 | XP_011955827.2 | 2 - 3 | 434 | 60 | ||
GenBank mRNA | X55152.1 | - | 394 | 60 | ||
X55966.1 | - | 238 | 60 | |||
X56756.1 | - | 410 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
