Assay Details
Gene Symbol: LPAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009332.1 | NP_001009332.1 | 3 - 4 | 1013 | 101 | |
XM_012111055.3 | XP_011966445.1 | 3 - 4 | 1111 | 101 | ||
XM_012111061.3 | XP_011966451.1 | 3 - 4 | 1090 | 101 | ||
XM_027963887.1 | XP_027819688.1 | 3 - 4 | 997 | 101 | ||
GenBank mRNA | U18405.1 | - | 3 - 4 | 1013 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
