Assay Details
Gene Symbol: SERPINA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009799.2 | NP_001009799.1 | 3 - 4 | 1069 | 99 | |
XM_012098288.2 | XP_011953678.2 | 4 - 5 | 1133 | 99 | ||
XM_012098289.3 | XP_011953679.2 | 4 - 5 | 1343 | 99 | ||
GenBank mRNA | JQ425036.1 | - | 4 - 5 | 1186 | 99 | |
JQ425037.1 | - | 3 - 4 | 915 | 99 | ||
JQ425038.1 | - | 2 - 3 | 271 | 99 | ||
X15555.1 | - | 1070 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
