Assay Details
Gene Symbol: SOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280703.1 | NP_001267632.1 | 3 - 4 | 400 | 82 | |
GenBank mRNA | AY221462.1 | - | 2 - 3 | 196 | 82 | |
GQ221055.1 | - | 3 - 4 | 400 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
