Assay Details
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009420.2 | NP_001009420.1 | 2 - 3 | 235 | 84 | |
GenBank mRNA | Z18291.1 | - | 235 | 84 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009420.2 | NP_001009420.1 | 2 - 3 | 235 | 84 | |
GenBank mRNA | Z18291.1 | - | 235 | 84 |