Assay Details
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009420.2 | NP_001009420.1 | 4 - 5 | 366 | 77 | |
GenBank mRNA | Z18291.1 | - | 366 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
