Assay Details
Gene Symbol: MADCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038020.1 | NP_001033109.1 | 1 - 2 | 81 | 114 | |
GenBank mRNA | DQ288287.1 | - | 1 - 2 | 81 | 114 |

More Information
Set Membership: |
![]() ![]() |
