Assay Details
Gene Symbol: IGFBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129733.1 | NP_001123205.1 | 1 - 2 | 346 | 89 | |
XM_012110889.3 | XP_011966279.1 | 1 - 2 | 1262 | 89 | ||
GenBank mRNA | EU727460.1 | - | 1 - 2 | 346 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
