Assay Details
Gene Symbol: MYD88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166183.1 | NP_001159655.1 | 3 - 4 | 644 | 61 | |
GenBank mRNA | GQ221044.1 | - | 3 - 4 | 644 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
