Assay Details
Gene Symbol: AHSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009802.1 | NP_001009802.1 | 3 - 4 | 464 | 114 | |
GenBank mRNA | X16578.1 | - | 464 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
