Assay Details
Gene Symbol: SLC25A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142890.1 | NP_001136362.1 | 3 - 4 | 368 | 64 | |
XM_012185066.3 | XP_012040456.2 | 4 - 5 | 472 | 64 | ||
XM_012185067.3 | XP_012040457.2 | 5 - 6 | 547 | 64 | ||
XM_012185068.3 | XP_012040458.2 | 5 - 6 | 519 | 64 | ||
XM_012185069.3 | XP_012040459.2 | 4 - 5 | 444 | 64 | ||
GenBank mRNA | FJ422548.1 | - | 3 - 4 | 368 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
