Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009426.1 | NP_001009426.1 | 2 - 3 | 959 | 90 | |
GenBank mRNA | Z68501.1 | - | 2 - 3 | 959 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
