Assay Details
Gene Symbol: RABAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161869.1 | NP_001155341.1 | 1 - 2 | 55 | 86 | |
GenBank mRNA | FJ943977.1 | - | 1 - 2 | 55 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
